Assay Details
| Gene Symbol: Ghrl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286404.1 | NP_001273333.1 | 3 - 4 | 356 | 104 | |
| NM_001286406.1 | NP_001273335.1 | 2 - 3 | 242 | 104 | ||
| NM_021488.5 | NP_067463.2 | 4 - 5 | 407 | 104 | ||
| GenBank mRNA | AB035701.1 | - | 3 - 4 | 349 | 104 | |
| AJ243503.1 | - | 3 - 4 | 372 | 104 | ||
| AK008658.1 | - | 4 - 5 | 377 | 104 | ||
| AK008860.1 | - | 4 - 5 | 377 | 104 | ||
| BC132230.1 | - | 3 - 4 | 373 | 40130507 | 104 | |
| BC132232.1 | - | 3 - 4 | 373 | 40130509 | 104 | |
| FJ914266.1 | - | 3 - 4 | 413 | 104 | ||
More Information
Set Membership: |
|

Ghrl
Genomic Map