Assay Details
| Gene Symbol: Fgf11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291104.1 | NP_001278033.1 | 4 - 5 | 613 | 75 | |
| NM_010198.2 | NP_034328.1 | 4 - 5 | 697 | 75 | ||
| GenBank mRNA | BC066859.1 | - | 4 - 5 | 613 | 30101881 | 75 |
| U66203.1 | - | 4 - 5 | 606 | 75 | ||
More Information
Set Membership: |
|

Fgf11
Genomic Map