Assay Details
| Gene Symbol: Prrg3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC060135.1 | - | 2 - 3 | 64 | 69 | |
| BC137616.1 | - | 2 - 3 | 192 | 8860635 | 70 | |
| BC137619.1 | - | 2 - 3 | 192 | 8860638 | 70 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC137616.1, BC137619.1 transcripts.
Set Membership: |
|

Prrg3
Genomic Map