Assay Details
Gene Symbol: Spsb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF403037.1 | - | 1 - 2 | 662 | 96 | |
AI849155.1 | - | 1 - 2 | 367 | 96 | ||
AK036451.1 | - | 1 - 2 | 202 | 96 | ||
AK148177.1 | - | 1 - 2 | 758 | 96 | ||
AK209543.1 | - | 1 - 2 | 109 | 96 | ||
BC002005.1 | - | 2 - 3 | 846 | 3496382 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
