Assay Details
| Gene Symbol: Aebp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK155272.1 | - | 8 - 9 | 964 | 119 | |
| BC057681.1 | - | 8 - 9 | 1012 | 3499299 | 119 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Aebp2| Gene Symbol: Aebp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK155272.1 | - | 8 - 9 | 964 | 119 | |
| BC057681.1 | - | 8 - 9 | 1012 | 3499299 | 119 | |
Set Membership: |
|