Assay Details
| Gene Symbol: Cldn34c1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK014019.1 | - | 4 | 891 | 71 | |
| AK032219.1 | - | 3 | 918 | 71 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cldn34c1| Gene Symbol: Cldn34c1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK014019.1 | - | 4 | 891 | 71 | |
| AK032219.1 | - | 3 | 918 | 71 | ||
Set Membership: |
|