Assay Details
Gene Symbol: Tmem171 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025606.1 | NP_001020777.1 | 3 - 4 | 880 | 62 | |
GenBank mRNA | BC099490.1 | - | 3 - 4 | 880 | 30522938 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
