Assay Details
| Gene Symbol: Rprml | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033212.2 | NP_001028384.1 | 1 | 423 | 94 | |
| GenBank mRNA | AK141598.1 | - | 1 | 424 | 94 | |
| BC147064.1 | - | 1 | 398 | 9056034 | 94 | |
| BC147065.1 | - | 1 | 398 | 9056062 | 94 | |
More Information
Set Membership: |
|

Rprml
Genomic Map