Assay Details
| Gene Symbol: Alg10b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033441.3 | NP_001028613.1 | 1 - 2 | 299 | 59 | |
| GenBank mRNA | AK143038.1 | - | 1 - 2 | 305 | 59 | |
| AK147821.1 | - | 1 - 2 | 300 | 59 | ||
| AK153580.1 | - | 1 - 2 | 279 | 59 | ||
| AK166344.1 | - | 1 - 2 | 301 | 59 | ||
| AK170450.1 | - | 1 - 2 | 300 | 59 | ||
| AK171179.1 | - | 1 - 2 | 261 | 59 | ||
More Information
Set Membership: |
|

Alg10b
Genomic Map