Assay Details
| Gene Symbol: Alg10b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033441.3 | NP_001028613.1 | 2 - 3 | 495 | 95 | |
| GenBank mRNA | AK143038.1 | - | 2 - 3 | 501 | 95 | |
| AK147821.1 | - | 2 - 3 | 496 | 95 | ||
| AK166344.1 | - | 2 - 3 | 497 | 95 | ||
| AK170450.1 | - | 2 - 3 | 496 | 95 | ||
More Information
Set Membership: |
|

Alg10b
Genomic Map