Assay Details
| Gene Symbol: Prex2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033636.4 | NP_001028808.1 | 4 - 5 | 493 | 75 | |
| NM_029525.1 | NP_083801.1 | 38 - 39 | 5010 | 75 | ||
| GenBank mRNA | AK136059.1 | - | 4 - 5 | 494 | 75 | |
| AM109952.1 | - | 38 - 39 | 5011 | 75 | ||
| BC139253.1 | - | 4 - 5 | 453 | 75 | ||
| BC139255.1 | - | 4 - 5 | 453 | 75 | ||
More Information
Set Membership: |
|

Prex2
Genomic Map