Assay Details
| Gene Symbol: Tmcc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK145948.1 | - | 1 - 2 | 190 | 64 | |
| AK158436.1 | - | 1 - 2 | 191 | 64 | ||
| AK165309.1 | - | 1 - 2 | 190 | 64 | ||
| AK172313.1 | - | 1 - 2 | 190 | 64 | ||
| AK172382.1 | - | 1 - 2 | 190 | 64 | ||
| AK172541.1 | - | 1 - 2 | 190 | 64 | ||
| AK172720.1 | - | 1 - 2 | 190 | 64 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK158436.1, AK165309.1, AK172313.1, AK172541.1 transcripts.
Set Membership: |
|

Tmcc2
Genomic Map