Assay Details
| Gene Symbol: Timm17b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011591.4 | NP_035721.1 | 2 - 3 | 170 | 68 | |
| GenBank mRNA | AF106621.1 | - | 2 - 3 | 124 | 68 | |
| AK003204.1 | - | 2 - 3 | 143 | 68 | ||
| AK009164.1 | - | 2 - 3 | 172 | 68 | ||
| AK011830.1 | - | 2 - 3 | 171 | 68 | ||
| AK049584.1 | - | 2 - 3 | 171 | 68 | ||
| AK150836.1 | - | 2 - 3 | 173 | 68 | ||
| AK151396.1 | - | 2 - 3 | 178 | 68 | ||
| AK153097.1 | - | 2 - 3 | 171 | 68 | ||
| BC008275.1 | - | 2 - 3 | 152 | 2811777 | 68 | |
More Information
Set Membership: |
|

Timm17b
Genomic Map