Assay Details
| Gene Symbol: Zfand1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025512.2 | NP_079788.2 | 5 - 6 | 366 | 72 | |
| GenBank mRNA | AK009240.1 | - | 5 - 6 | 380 | 72 | |
| AK017696.1 | - | 5 - 6 | 380 | 72 | ||
| AK030472.1 | - | 5 - 6 | 374 | 72 | ||
| AK077873.1 | - | 5 - 6 | 376 | 72 | ||
| BC023352.1 | - | 5 - 6 | 358 | 72 | ||
| BC025216.1 | - | 5 - 6 | 358 | 72 | ||
| BC046585.1 | - | 5 - 6 | 366 | 6314252 | 72 | |
| BC089494.1 | - | 5 - 6 | 378 | 30504018 | 72 | |
More Information
Set Membership: |
|

Zfand1
Genomic Map