Assay Details
| Gene Symbol: Zfp354a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009329.3 | NP_033355.2 | 2 - 3 | 230 | 56 | |
| GenBank mRNA | AB221586.1 | - | 1 - 2 | 34 | 56 | |
| AF184111.1 | - | 2 - 3 | 192 | 56 | ||
| AK030469.1 | - | 2 - 3 | 208 | 56 | ||
| BC050843.1 | - | 2 - 3 | 230 | 6506256 | 56 | |
| L77247.1 | - | 1 - 2 | 94 | 56 | ||
More Information
Set Membership: |
|

Zfp354a
Genomic Map