Assay Details
| Gene Symbol: Calm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007589.5 | NP_031615.1 | 3 - 4 | 316 | 80 | |
| GenBank mRNA | AK012247.1 | - | 3 - 4 | 244 | 80 | |
| AK012564.1 | - | 3 - 4 | 245 | 80 | ||
| AK013068.1 | - | 3 - 4 | 238 | 80 | ||
| AK152303.1 | - | 3 - 4 | 248 | 80 | ||
| AK160636.1 | - | 3 - 4 | 251 | 80 | ||
| AK161268.1 | - | 3 - 4 | 244 | 80 | ||
| AK161984.1 | - | 3 - 4 | 245 | 80 | ||
| AK168741.1 | - | 3 - 4 | 245 | 80 | ||
| BC021347.1 | - | 3 - 4 | 247 | 4006686 | 80 | |
| BC100301.1 | - | 3 - 4 | 231 | 30526959 | 80 | |
| L31642.1 | - | 3 - 4 | 251 | 80 | ||
More Information
Set Membership: |
|

Calm2
Genomic Map