Assay Details
| Gene Symbol: Calm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007589.5 | NP_031615.1 | 5 - 6 | 559 | 80 | |
| GenBank mRNA | AK002974.1 | - | 520 | 80 | ||
| AK012247.1 | - | 5 - 6 | 487 | 80 | ||
| AK012564.1 | - | 5 - 6 | 488 | 80 | ||
| AK013068.1 | - | 5 - 6 | 481 | 80 | ||
| AK152303.1 | - | 5 - 6 | 491 | 80 | ||
| AK160636.1 | - | 5 - 6 | 494 | 80 | ||
| AK161268.1 | - | 5 - 6 | 487 | 80 | ||
| AK161984.1 | - | 5 - 6 | 488 | 80 | ||
| AK168741.1 | - | 5 - 6 | 488 | 80 | ||
| BC010730.1 | - | 3 - 4 | 264 | 80 | ||
| BC021347.1 | - | 5 - 6 | 490 | 4006686 | 80 | |
| BC051444.1 | - | 5 - 6 | 483 | 30072702 | 80 | |
| BC100301.1 | - | 5 - 6 | 474 | 30526959 | 80 | |
More Information
Set Membership: |
|

Calm2
Genomic Map