Assay Details
Gene Symbol: Cort | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007745.3 | NP_031771.1 | 1 - 2 | 209 | 82 | |
GenBank mRNA | AF013253.1 | - | 1 - 2 | 130 | 82 | |
AK013496.1 | - | 1 - 2 | 211 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
