Assay Details
| Gene Symbol: Gas1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008086.2 | NP_032112.1 | 1 | 1516 | 94 | |
| GenBank mRNA | BC055747.1 | - | 2 | 611 | 94 | |
| BC058628.1 | - | 2 | 1156 | 6401957 | 94 | |
| X65128.1 | - | 1 | 1346 | 94 | ||
More Information
Set Membership: |
|

Gas1
Genomic Map