Assay Details
| Gene Symbol: Hccs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008222.4 | NP_032248.3 | 1 - 2 | 228 | 118 | |
| GenBank mRNA | AK077566.1 | - | 1 - 2 | 166 | 118 | |
| AK143150.1 | - | 1 - 2 | 234 | 119 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK077566.1, NM_008222.4 transcripts.
Set Membership: |
|

Hccs
Genomic Map