Assay Details
Gene Symbol: Hoxa10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008263.3 | NP_032289.2 | 1 - 2 | 985 | 60 | |
GenBank mRNA | BC050839.1 | - | 1 - 2 | 957 | 6511608 | 60 |
L08757.1 | - | 1 - 2 | 985 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
