Assay Details
| Gene Symbol: Hspe1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008303.4 | NP_032329.1 | 3 - 4 | 517 | 96 | |
| GenBank mRNA | AK088121.1 | - | 3 - 4 | 348 | 96 | |
| AK133876.1 | - | 3 - 4 | 324 | 96 | ||
| AK151935.1 | - | 3 - 4 | 327 | 96 | ||
| BC024385.1 | - | 3 - 4 | 276 | 5004427 | 96 | |
| BC099385.1 | - | 3 - 4 | 328 | 30848267 | 96 | |
| CJ238411.1 | - | 2 - 3 | 201 | 96 | ||
| U09659.1 | - | 3 - 4 | 333 | 96 | ||
More Information
Set Membership: |
|

Hspe1
Genomic Map