Assay Details
| Gene Symbol: Ndrg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008681.2 | NP_032707.2 | 10 - 11 | 855 | 77 | |
| GenBank mRNA | AA795988.1 | - | 2 - 3 | 148 | 77 | |
| AK004548.1 | - | 10 - 11 | 855 | 77 | ||
| AK161860.1 | - | 10 - 11 | 857 | 77 | ||
| AK165007.1 | - | 10 - 11 | 855 | 77 | ||
| AK170281.1 | - | 10 - 11 | 856 | 77 | ||
| AK170636.1 | - | 10 - 11 | 854 | 77 | ||
| AK170809.1 | - | 10 - 11 | 855 | 77 | ||
| BC015282.1 | - | 10 - 11 | 810 | 4237943 | 77 | |
| BC071235.1 | - | 10 - 11 | 863 | 4242075 | 77 | |
| BC141246.1 | - | 10 - 11 | 746 | 9055858 | 77 | |
| BC171968.1 | - | 10 - 11 | 746 | 9054254 | 77 | |
| CA320478.1 | - | 7 - 8 | 598 | 77 | ||
| U60593.1 | - | 10 - 11 | 853 | 77 | ||
More Information
Set Membership: |
|

Ndrg1
Genomic Map