Assay Details
| Gene Symbol: Ndrg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008681.2 | NP_032707.2 | 3 - 4 | 255 | 62 | |
| GenBank mRNA | AK004548.1 | - | 3 - 4 | 255 | 62 | |
| AK098075.1 | - | 3 - 4 | 254 | 62 | ||
| AK165007.1 | - | 3 - 4 | 255 | 62 | ||
| AK170281.1 | - | 3 - 4 | 256 | 62 | ||
| AK170439.1 | - | 3 - 4 | 254 | 62 | ||
| AK170636.1 | - | 3 - 4 | 254 | 62 | ||
| AK170809.1 | - | 3 - 4 | 255 | 62 | ||
| AK199494.1 | - | 2 - 3 | 79 | 62 | ||
| AW611038.1 | - | 3 - 4 | 255 | 62 | ||
| BC015282.1 | - | 3 - 4 | 210 | 4237943 | 62 | |
| BC071235.1 | - | 3 - 4 | 263 | 4242075 | 62 | |
| BC141246.1 | - | 3 - 4 | 146 | 9055858 | 62 | |
| BC171968.1 | - | 3 - 4 | 146 | 9054254 | 62 | |
| BY232155.1 | - | 3 - 4 | 256 | 62 | ||
| U60593.1 | - | 3 - 4 | 253 | 62 | ||
More Information
Set Membership: |
|

Ndrg1
Genomic Map