Assay Details
| Gene Symbol: Rps29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009093.2 | NP_033119.1 | 1 - 2 | 195 | 84 | |
| GenBank mRNA | AA217935.1 | - | 1 - 2 | 79 | 84 | |
| AK002939.1 | - | 1 - 2 | 99 | 84 | ||
| AK012285.1 | - | 1 - 2 | 98 | 84 | ||
| AK224925.1 | - | 1 - 2 | 88 | 84 | ||
| BC024393.1 | - | 115 | 4986748 | 84 | ||
| BC051203.1 | - | 1 - 2 | 82 | 1476903 | 84 | |
| L31609.1 | - | 1 - 2 | 79 | 84 | ||
More Information
Set Membership: |
|

Rps29
Genomic Map