Assay Details
Gene Symbol: Olfr1357 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011737.2 | NP_001011737.1 | 1 - 2 | 174 | 108 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001011737.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
