Assay Details
| Gene Symbol: Sat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291865.1 | NP_001278794.1 | 5 - 6 | 693 | 108 | |
| NM_009121.4 | NP_033147.1 | 4 - 5 | 583 | 108 | ||
| GenBank mRNA | AK002531.1 | - | 4 - 5 | 484 | 108 | |
| AK131657.1 | - | 1 - 2 | 870 | 108 | ||
| AK154445.1 | - | 4 - 5 | 479 | 108 | ||
| BC058696.1 | - | 4 - 5 | 543 | 6820169 | 108 | |
| BC108334.1 | - | 4 - 5 | 894 | 108 | ||
| L10244.1 | - | 4 - 5 | 391 | 108 | ||
More Information
Set Membership: |
|

Sat1
Genomic Map