Assay Details
| Gene Symbol: Sox15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009235.2 | NP_033261.1 | 1 - 2 | 856 | 86 | |
| GenBank mRNA | AF182945.1 | - | 1 - 2 | 520 | 86 | |
| AK131957.1 | - | 1 - 2 | 858 | 86 | ||
| BC119067.1 | - | 1 - 2 | 580 | 8733816 | 86 | |
| BC119069.1 | - | 1 - 2 | 580 | 8733818 | 86 | |
| X98369.1 | - | 1 - 2 | 247 | 86 | ||
More Information
Set Membership: |
|

Sox15
Genomic Map