Assay Details
| Gene Symbol: Pglyrp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009402.2 | NP_033428.1 | 1 - 2 | 289 | 68 | |
| GenBank mRNA | AF076482.1 | - | 1 - 2 | 288 | 68 | |
| AF193843.1 | - | 1 - 2 | 277 | 68 | ||
| AK008335.1 | - | 1 - 2 | 290 | 68 | ||
| BC005582.1 | - | 1 - 2 | 299 | 3969014 | 68 | |
| CT010165.1 | - | 1 - 2 | 253 | 68 | ||
| X86374.1 | - | 1 - 2 | 288 | 68 | ||
More Information
Set Membership: |
|

Pglyrp1
Genomic Map