Assay Details
| Gene Symbol: Aoc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009675.2 | NP_033805.1 | 3 - 4 | 2351 | 99 | |
| GenBank mRNA | AF054831.1 | - | 3 - 4 | 2366 | 99 | |
| AF115411.1 | - | 3 - 4 | 2182 | 99 | ||
| AK040729.1 | - | 1178 | 99 | |||
| BC080857.1 | - | 3 - 4 | 2197 | 6854465 | 99 | |
More Information
Set Membership: |
|

Aoc3
Genomic Map