Assay Details
| Gene Symbol: Capza1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009797.2 | NP_033927.2 | 7 - 8 | 712 | 97 | |
| GenBank mRNA | AK146296.1 | - | 7 - 8 | 598 | 97 | |
| AK147049.1 | - | 7 - 8 | 601 | 97 | ||
| AK149602.1 | - | 7 - 8 | 598 | 97 | ||
| AK151253.1 | - | 7 - 8 | 609 | 97 | ||
| AK152364.1 | - | 7 - 8 | 597 | 97 | ||
| AK161370.1 | - | 7 - 8 | 714 | 97 | ||
| AK163418.1 | - | 7 - 8 | 598 | 97 | ||
| AK165655.1 | - | 7 - 8 | 600 | 97 | ||
| AK166484.1 | - | 7 - 8 | 596 | 97 | ||
| AK166964.1 | - | 7 - 8 | 601 | 97 | ||
| AK167193.1 | - | 7 - 8 | 599 | 97 | ||
| AK168099.1 | - | 7 - 8 | 598 | 97 | ||
| AK168916.1 | - | 7 - 8 | 600 | 97 | ||
| AK169806.1 | - | 7 - 8 | 598 | 97 | ||
| BC016232.1 | - | 7 - 8 | 601 | 4489604 | 97 | |
| BC085506.1 | - | 7 - 8 | 587 | 6817308 | 97 | |
| U16740.1 | - | 7 - 8 | 581 | 97 | ||
More Information
Set Membership: |
|

Capza1
Genomic Map