Assay Details
| Gene Symbol: Khsrp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010613.3 | NP_034743.3 | 11 - 12 | 1189 | 108 | |
| GenBank mRNA | AK052249.1 | - | 7 - 8 | 640 | 108 | |
| AK077415.1 | - | 11 - 12 | 1159 | 108 | ||
| AK156541.1 | - | 11 - 12 | 1167 | 108 | ||
| BC108414.1 | - | 5 - 6 | 490 | 108 | ||
More Information
Set Membership: |
|

Khsrp
Genomic Map