Assay Details
| Gene Symbol: Khsrp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010613.3 | NP_034743.3 | 13 - 14 | 1593 | 74 | |
| GenBank mRNA | AK018152.1 | - | 1 - 2 | 52 | 74 | |
| AK052249.1 | - | 9 - 10 | 1045 | 74 | ||
| AK077415.1 | - | 13 - 14 | 1563 | 74 | ||
| AK156541.1 | - | 13 - 14 | 1571 | 74 | ||
| BC064454.1 | - | 2 - 3 | 174 | 74 | ||
| BC108414.1 | - | 7 - 8 | 894 | 74 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK018152.1 transcripts.
Set Membership: |
|

Khsrp
Genomic Map