Assay Details
| Gene Symbol: Khsrp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010613.3 | NP_034743.3 | 16 - 17 | 2006 | 90 | |
| GenBank mRNA | AK018152.1 | - | 4 - 5 | 464 | 90 | |
| AK052249.1 | - | 12 - 13 | 1458 | 90 | ||
| AK077415.1 | - | 16 - 17 | 1976 | 90 | ||
| AK156541.1 | - | 16 - 17 | 1984 | 90 | ||
| BC064454.1 | - | 5 - 6 | 587 | 90 | ||
| BC108414.1 | - | 10 - 11 | 1307 | 90 | ||
More Information
Set Membership: |
|

Khsrp
Genomic Map