Assay Details
| Gene Symbol: Khsrp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010613.3 | NP_034743.3 | 8 - 9 | 893 | 71 | |
| GenBank mRNA | AK052249.1 | - | 4 - 5 | 344 | 71 | |
| AK077415.1 | - | 8 - 9 | 863 | 71 | ||
| AK156541.1 | - | 8 - 9 | 871 | 71 | ||
| AK203432.1 | - | 2 - 3 | 244 | 71 | ||
| BC108414.1 | - | 2 - 3 | 194 | 71 | ||
| CB247078.1 | - | 7 - 8 | 466 | 71 | ||
More Information
Set Membership: |
|

Khsrp
Genomic Map