Assay Details
| Gene Symbol: Lbx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010691.5 | NP_034821.2 | 1 - 2 | 340 | 87 | |
| GenBank mRNA | AB108497.1 | - | 1 - 2 | 340 | 87 | |
| AK144562.1 | - | 1 - 2 | 185 | 87 | ||
| BC119177.1 | - | 1 - 2 | 429 | 8733926 | 87 | |
| BC120586.1 | - | 1 - 2 | 429 | 40129509 | 87 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK144562.1 transcripts.
Set Membership: |
|

Lbx1
Genomic Map