Assay Details
| Gene Symbol: Rbmy | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011253.2 | NP_035383.2 | 2 - 3 | 219 | 108 | |
| GenBank mRNA | Y15131.1 | - | 2 - 3 | 208 | 108 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_011253.2, Y15131.1 transcripts.
Set Membership: |
|

Rbmy
Genomic Map