Assay Details
| Gene Symbol: Fabp9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011598.3 | NP_035728.2 | 1 - 2 | 190 | 83 | |
| GenBank mRNA | AK005745.1 | - | 1 - 2 | 134 | 83 | |
| AK161314.1 | - | 1 - 2 | 95 | 83 | ||
| BC048437.1 | - | 1 - 2 | 129 | 6773242 | 83 | |
More Information
Set Membership: |
|

Fabp9
Genomic Map