Assay Details
| Gene Symbol: Vbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011692.2 | NP_035822.2 | 1 - 2 | 129 | 67 | |
| GenBank mRNA | AK002282.1 | - | 1 - 2 | 99 | 67 | |
| AK008847.1 | - | 1 - 2 | 99 | 67 | ||
| AK077923.1 | - | 1 - 2 | 114 | 67 | ||
| AK151527.1 | - | 1 - 2 | 99 | 67 | ||
| AK153069.1 | - | 1 - 2 | 98 | 67 | ||
| AK159284.1 | - | 1 - 2 | 99 | 67 | ||
| AK167064.1 | - | 1 - 2 | 99 | 67 | ||
| AK167742.1 | - | 1 - 2 | 130 | 67 | ||
| AK169185.1 | - | 1 - 2 | 99 | 67 | ||
| BC132169.1 | - | 1 - 2 | 72 | 67 | ||
| BC132171.1 | - | 1 - 2 | 72 | 67 | ||
| BY134471.1 | - | 1 - 2 | 122 | 67 | ||
| DV066998.1 | - | 1 - 2 | 162 | 67 | ||
More Information
Set Membership: |
|

Vbp1
Genomic Map