Assay Details
| Gene Symbol: Vbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011692.2 | NP_035822.2 | 5 - 6 | 556 | 122 | |
| GenBank mRNA | AK002282.1 | - | 5 - 6 | 526 | 122 | |
| AK008847.1 | - | 5 - 6 | 526 | 122 | ||
| AK077923.1 | - | 5 - 6 | 541 | 122 | ||
| AK151527.1 | - | 5 - 6 | 526 | 122 | ||
| AK153069.1 | - | 5 - 6 | 525 | 122 | ||
| AK159284.1 | - | 5 - 6 | 526 | 122 | ||
| AK167064.1 | - | 5 - 6 | 526 | 122 | ||
| AK169185.1 | - | 5 - 6 | 526 | 122 | ||
| BC132169.1 | - | 5 - 6 | 499 | 122 | ||
| BC132171.1 | - | 5 - 6 | 499 | 122 | ||
| DV066998.1 | - | 5 - 6 | 589 | 122 | ||
| U96760.1 | - | 5 - 6 | 507 | 122 | ||
More Information
Set Membership: |
|

Vbp1
Genomic Map