Assay Details
Gene Symbol: Mt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013603.2 | NP_038631.1 | 2 - 3 | 284 | 79 | |
GenBank mRNA | AK140209.1 | - | 2 - 3 | 285 | 79 | |
BC059725.1 | - | 2 - 3 | 182 | 6600430 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
