Assay Details
| Gene Symbol: Hunk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015755.2 | NP_056570.1 | 6 - 7 | 1068 | 58 | |
| GenBank mRNA | AF055919.2 | - | 6 - 7 | 1068 | 58 | |
| AF167987.1 | - | 6 - 7 | 1080 | 58 | ||
| AK047337.1 | - | 2 - 3 | 2507 | 58 | ||
| AK144639.1 | - | 6 - 7 | 1051 | 58 | ||
| AK147425.1 | - | 6 - 7 | 1453 | 58 | ||
| AK147734.1 | - | 6 - 7 | 822 | 58 | ||
| BC137788.1 | - | 6 - 7 | 1129 | 8860807 | 58 | |
| BC137790.1 | - | 6 - 7 | 1128 | 8860809 | 58 | |
More Information
Set Membership: |
|

Hunk
Genomic Map