Assay Details
| Gene Symbol: B3gnt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016888.5 | NP_058584.3 | 1 - 2 | 245 | 98 | |
| GenBank mRNA | AK160088.1 | - | 1 - 2 | 214 | 98 | |
| AY043479.1 | - | 1 - 2 | 203 | 98 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK160088.1, AY043479.1, NM_016888.5 transcripts.
Set Membership: |
|

B3gnt2
Genomic Map