Assay Details
| Gene Symbol: Tlr5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016928.2 | NP_058624.2 | 3 - 4 | 323 | 86 | |
| GenBank mRNA | AK036842.1 | - | 3 - 4 | 325 | 86 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK036842.1, NM_016928.2 transcripts.
Set Membership: |
|

Tlr5
Genomic Map