Assay Details
| Gene Symbol: Sfn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018754.2 | NP_061224.2 | 1 | 1019 | 96 | |
| GenBank mRNA | AF058798.1 | - | 1 | 783 | 96 | |
| AF152893.1 | - | 1 | 620 | 96 | ||
| AK146490.1 | - | 1 | 693 | 96 | ||
| AK169358.1 | - | 1 | 684 | 96 | ||
More Information
Set Membership: |
|

Sfn
Genomic Map