Assay Details
| Gene Symbol: Olfr157 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019475.3 | NP_062348.1 | 1 | 1221 | 109 | |
| GenBank mRNA | BC137911.1 | - | 1 | 1218 | 8860930 | 109 | 
| BC137912.1 | - | 1 | 1218 | 8860931 | 109 | |
More Information
Set Membership: | 
		   
					
						
							 | 
				

Olfr157
Genomic Map