Assay Details
| Gene Symbol: Metap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019648.3 | NP_062622.1 | 3 - 4 | 958 | 88 | |
| GenBank mRNA | AB003144.1 | - | 2 - 3 | 269 | 88 | |
| AF434712.1 | - | 2 - 3 | 348 | 88 | ||
| AK076804.1 | - | 2 - 3 | 301 | 88 | ||
| AK090323.1 | - | 2 - 3 | 289 | 88 | ||
| AK147097.1 | - | 2 - 3 | 363 | 88 | ||
| AK159650.1 | - | 2 - 3 | 373 | 88 | ||
| AK163318.1 | - | 2 - 3 | 375 | 88 | ||
| AK165301.1 | - | 3 - 4 | 960 | 88 | ||
| AK165845.1 | - | 2 - 3 | 353 | 88 | ||
| AK189664.1 | - | 1 - 2 | 87 | 88 | ||
| BC002213.1 | - | 2 - 3 | 370 | 3490341 | 88 | |
| BC010263.1 | - | 2 - 3 | 343 | 88 | ||
| BC092052.1 | - | 2 - 3 | 335 | 3670685 | 88 | |
More Information
Set Membership: |
|

Metap2
Genomic Map