Assay Details
| Gene Symbol: Akap10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019921.3 | NP_064305.2 | 3 - 4 | 506 | 71 | |
| GenBank mRNA | AK049399.1 | - | 3 - 4 | 475 | 71 | |
| AK051044.1 | - | 1 - 2 | 141 | 71 | ||
| AK083255.1 | - | 3 - 4 | 475 | 71 | ||
| AK162966.1 | - | 3 - 4 | 507 | 71 | ||
More Information
Set Membership: |
|

Akap10
Genomic Map