Assay Details
| Gene Symbol: Mos | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020021.2 | NP_064405.2 | 1 | 716 | 76 | |
| GenBank mRNA | AK133089.1 | - | 1 | 716 | 76 | |
| BC137690.1 | - | 1 | 629 | 8860709 | 76 | |
| BC145745.1 | - | 1 | 629 | 40131048 | 76 | |
More Information
Set Membership: |
|

Mos
Genomic Map