Assay Details
| Gene Symbol: Ndnl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023239.4 | NP_075728.1 | 1 | 617 | 81 | |
| GenBank mRNA | AF319979.1 | - | 1 | 495 | 81 | |
| AK010294.1 | - | 1 | 578 | 81 | ||
| AK017727.1 | - | 1 | 619 | 81 | ||
| AK018250.1 | - | 1 | 578 | 81 | ||
| AK049759.1 | - | 1 | 597 | 81 | ||
| AK076471.1 | - | 1 | 603 | 81 | ||
| AK131157.1 | - | 574 | 81 | |||
| BC034892.1 | - | 1 | 542 | 1494952 | 81 | |
| BC092289.1 | - | 1 | 579 | 4990134 | 81 | |
More Information
Set Membership: |
|

Ndnl2
Genomic Map